missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PGD2 Synthase/PTGDS Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
Description
A peptide to PTGDS.
Specifications
Specifications
| Gene ID (Entrez) | 5730 |
| Species | Human |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB110-59911. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | Beta-trace protein, cerebrin-28, EC 5.3.99.2, Glutathione-independent PGD synthase, glutathione-independent PGD synthetase, lipocalin-type prostaglandin D synthase, Lipocalin-type prostaglandin-D synthase, L-PGDS, PDS, PGD2, PGD2 synthase, PGDS2, PGDSLPGDS, prostaglandin D synthase, prostaglandin D2 synthase (21 kD, brain), prostaglandin D2 synthase 21 kDa (brain), Prostaglandin-D2 synthase, prostaglandin-H2 D-isomerase |
| Gene Symbol | PTGDS |
| Product Type | PGD2 Synthase/PTGDS |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction