missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ nfnB Protein

Product Code. 18225354 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.10mg
0.50mg
This item is not returnable. View return policy

Product Code. 18225354

Brand: Novus Biologicals™ NBP1460350.1MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-217 of E.coli nfnB The Recombinant E. coli nfnB Protein is derived from E. coli. The Recombinant E. coli nfnB Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.0mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation In 20mM Tris-HCl buffer (pH 8.0) containing 50mM NaCl, 1mM DTT, 10% glycerol
Molecular Weight (g/mol) 26.0kDa
Purification Method Protein
Quantity 0.1 mg
Source Bacteria
Immunogen MGSSHHHHHH SSGLVPRGSH MDIISVALKR HSTKAFDASK KLTPEQAEQI KTLLQYSPSS TNSQPWHFIV ASTEEGKARV AKSAAGNYVF NERKMLDASH VVVFCAKTAM DDVWLKLVVD QEDADGRFAT PEAKAANDKG RKFFADMHRK DLHDDAEWMA KQVYLNVGNF LLGVAALGLD AVPIEGFDAA ILDAEFGLKE KGYTSLVVVP VGHHSVEDFN ATLPKSRLPQ NITLTEV
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name nfnB
Conjugate Unconjugated
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.