missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Melatonin Receptor 1B Antibody Blocking Peptide

Product Code. 18374654 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374654 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18374654 Supplier Novus Biologicals™ Supplier No. NLS932PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

The Melatonin Receptor 1B Blocking Peptide has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 4544
Species Human
Accession Number P49286
Content And Storage Store at -80 C. Avoid freeze-thaw cycles.
Formulation PBS, pH 7.4
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NLS932. To Block The Specific Antibody Activity in Immunohistochemistry Analysis, Pre-Incubate The Antibody with a 10-100 x Molar Excess of Peptide for 60 Minutes (at Room Temperature. Optimization of The Molar Ratio of Antigen to Antibody May be Required. Centrifugation of The Pre-Adsorbed Solution Prior to Addition to The Slide May Decrease Background Due to Non-Specific Binding of Large Antibody-Antigen Complexes. For Further Blocking Peptide Related Protocol, Click Here .)
Gene Alias FGQTL2, Mel 1 b melatonin receptor, Mel1b receptor, MEL-1 B-R, melatonin receptor 1 B, melatonin receptor MEL1B, melatonin receptor type 1 B, MT2
Gene Symbol MTNR1B
Product Type Melatonin Receptor 1B
Quantity 0.05 mg
Regulatory Status RUO - research use only
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.