missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MAT1/2A Antibody Blocking Peptide

Product Code. 18272320 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18272320 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18272320 Supplier Novus Biologicals™ Supplier No. NB11094162PEP
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A peptide to MAT2A.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 4144
Species Human
Content And Storage Store at -80 C. Avoid freeze-thaw cycles.
Formulation Peptide dissolved in dH2O. Contains no BSA.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB 110-94162. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias AdoMet synthase 2, AMS2, EC 2.5.1.6, MAT 2, MATA2SAMS2adoMet synthetase 2, MATII, MAT-II, Methionine adenosyltransferase 2, Methionine adenosyltransferase II, methionine adenosyltransferase II, alpha, S-adenosylmethionine synthase isoform type-2, S-adenosylmethionine synthetase isoform type-2
Gene Symbol MAT2A
Product Type MAT1/2A
Quantity 0.05 mg
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.