missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Kinesin 5A Antibody Blocking Peptide

Product Code. 18288785 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
This item is not returnable. View return policy

Product Code. 18288785

Brand: Novus Biologicals™ NBP176816PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide containing 20 amino acids near the carboxy terminus of carboxy terminus of human KIF5. Source: Synthetic (Accession #: Q12840) The Kinesin 5A Blocking Peptide has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 3798
Species Human
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-76816. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias D12S1889, KIF5A variant protein, kinesin family member 5 A, kinesin heavy chain isoform 5 A, Kinesin heavy chain neuron-specific 1, kinesin, heavy chain, neuron-specific, MY050, Neuronal kinesin heavy chain, NKHC1, NKHCspastic paraplegia 10 (autosomal dominant), SPG10
Gene Symbol KIF5A
Product Type Kinesin 5A
Quantity 0.05 mg
Regulatory Status RUO - research use only
Source Synthetic
Termination 20 amino acids near the carboxy terminus of human KIF5
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.