missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Jurkat Membrane Cell Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
Description
The Jurkat Membrane Cell Lysate is derived from human.
Specifications
Specifications
| Quantity | 0.1 mg |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Membrane Protein - Human Tumor Cell Line: Jurkat |
| Cellular Fraction | Membrane |
| Host Species | Human |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction