missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Imidazoquinoline Resiquimod (R-848), TLR7 and TLR8 Ligand

Product Code. 18735103 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18735103 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18735103 Supplier Novus Biologicals™ Supplier No. NBP226231

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Applications: Functional

Imidazoquinoline Resiquimod (R-848), TLR7 and TLR8 ligand is a low molecular weight synthetic molecule of the Imidazoquinoline family. It has potent immunostimulatory activity. It stimulates antibody secretion, cytokine production, protection from apoptosis and CD80 up-regulation.

  • Species: Human
  • Stimulation of TLR8 has been achieved with 0.5-5μg/mL

Recommended Storage

Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.

TRUSTED_SUSTAINABILITY

Specifications

Buffer DMSO
Content And Storage Store at -20°C.
For Use With (Application) Identity determined by MS, 1H-NMR and 13C-NMR. This product is useful for activation of mouse TLR7 and human TLR7 and TLR8. Stimulation of TLR8 has been achieved with 0.5-5 ug/mL. Use in functional assays reported in scientific literature (PMID 26134251).
Product Type Imidazoquinoline Resiquimod (R-848), TLR7 and TLR8 Ligand
Purity >98%, by HPLC.
Quantity 0.5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.