missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human ZNF514 His Protein

Product Code. 18116931 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18116931

Brand: Novus Biologicals™ NBP222957

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-400 of Human ZNF514 The Recombinant Human ZNF514 Protein is derived from E. coli. The Recombinant Human ZNF514 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_116177
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 84874
Molecular Weight (g/mol) 48.3kDa
Name ZNF514 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen ZNF514, 1-400aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMTFEDVA VEFSQWEWGQ LNPAQKDLYR EVMLENFRNL AILGLLVSKP YVICQLEEGG EPFMVEREIS TGAHSDWKRR SKSKESMPSW GISKEELFQV VSVEKHIQDV LQFSKLKAAC GCDGQLEMQQ IKQERHLKQM STIHKSATTL SRDYKWNGFG RSLGLRSVLV NQHSILMGEG SYKCDTEFRQ TLGGNNSQRT HPEKKSCKCN ECGKSFHFQS ELRRHQRCHT GEKPYECSDC GRAFGHISSL IKHQRTHTGE KPYECSECGR AFSQSSSLVL HYRFHTGEKP YKCNECGRAF GHTSSLIKHQ RTHTGEKPYE CRECGRTFSQ SSSLIVHYRF HTGEKPYKCN KCGRAFSQSS SLTQHYRFHT GEKPYKCNEC GRAFAHTASL IKHQRSHAGK KTL
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.