missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human SGTA His Protein
Shop All Bio Techne Products
Click to view available options
:
0.1mg; Unlabeled
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-313 of Human SGTA The Recombinant Human SGTA Protein is derived from E. coli. The Recombinant Human SGTA Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_003012 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT |
| Gene ID (Entrez) | 6449 |
| Molecular Weight (g/mol) | 35.1kDa |
| Name | SGTA Protein |
| Purification Method | Protein |
| Immunogen | MDNKKRLAYA IIQFLHDQLR HGGLSSDAQE SLEVAIQCLE TAFGVTVEDS DLALPQTLPE IFEAAATGKE MPQDLRSPAR TPPSEEDSAE AERLKTEGNE QMKVENFEAA VHFYGKAIEL NPANAVYFCN RAAAYSKLGN YAGAVQDCER AICIDPAYSK AYGRMGLALS SLNKHVEAVA YYKKALELDP DNETYKSNLK IAELKLREAP SPTGGVGSFD IAGLLNNPGF MSMASNLMNN PQIQQLMSGM ISGGNNPLGT PGTSPSQNDL ASLIQAGQQF AQQMQQQNPE LIEQLRSQIR SRTPSASNDD QQELEHHHHH H |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction