missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human S100P His Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-95 of Human S100P The Recombinant Human S100P Protein is derived from E. coli. The Recombinant Human S100P Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | P25815 |
| For Use With (Application) | Chromatin Immunoprecipitation, ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH8.0), 20% glycerol, 50mM NaCl |
| Gene ID (Entrez) | 6286 |
| Molecular Weight (g/mol) | 12.6kDa |
| Name | S100P Protein |
| Purification Method | Protein |
| Quantity | 0.1 mg |
| Source | E. Coli |
| Immunogen | S100P, 1-95aa, Human, His tag, E.coli (NP_005971). |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction