Learn More
Description
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 62 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
- Species: Human
- Cross reactivity: Human reactivity reported in scientific literature (PMID: 24865426)
- Tissue condition: Normal
- No. of samples: 49
- No. of patients: 40
- Core diameter: 2.0mm
- Section thickness: 4μm
Applications:
Immunohistochemistry-Paraffin
Recommended Storage
Store at 4°C. Do not freeze.
Services
1 Year
Specifications
Specifications
| Quantity | 1 Slide |
| Regulatory Status | RUO |
| Species | Human |
| Tissue Condition | Normal |
| Type | Prostate Cancer |
| For Use With (Application) | Immunohistochemistry-Paraffin, Hematoxylin & Eosin Stain |
| Cross Reactivity | Human reactivity reported in scientific literature (PMID: 24865426) |
This product is for research use only and is not approved for use in humans or in clinical diagnosis
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.