missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human PP2C gamma/PPM1G His Protein
Shop All Bio Techne Products
Click to view available options
:
0.1mg; Unlabeled
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 317-546 of Human PP2C gamma/PPM1G The Recombinant Human PP2C gamma/PPM1G Protein is derived from E. coli. The Recombinant Human PP2C gamma/PPM1G Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_002698 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 25mM Tris buffer (pH 7.5), 1mM DTT, 1mM EDTA, 2mM beta-mercaptoethanol, 20% glycerol |
| Gene ID (Entrez) | 5496 |
| Molecular Weight (g/mol) | 27kDa |
| Name | PP2C gamma/PPM1G Protein |
| Purification Method | Protein |
| Immunogen | MGSSHHHHHHSSGLVPRGSHMEGKEEPGSDSGTTAVVALIRGKQLIVANAGDSRCVVSEAGKALDMSYDHKPEDEVELARIKNAGGKVTMDGRVNGGLNLSRAIGDHFYKRNKNLPPEEQMISALPDIKVLTLTDDHEFMVIACDGIWNVMSSQEVVDFIQSKISQRDENGELRLLSSIVEELLDQCLAPDTSGDGTGCDNMTCIIICFKPRNTAELQPESGKRKLEEVLSTEGAEENGNSDKKKKAKRD |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction