missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human PARP2 His Protein

Product Code. 18150242 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18150242

Brand: Novus Biologicals™ NBP223339

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 233-583 of Human PARP2 The Recombinant Human PARP2 Protein is derived from E. coli. The Recombinant Human PARP2 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_005475
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 10038
Molecular Weight (g/mol) 42.5kDa
Name PARP2 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen PARP2, 233-583aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSHMQLDLR VQELIKLICN VQAMEEMMME MKYNTKKAPL GKLTVAQIKA GYQSLKKIED CIRAGQHGRA LMEACNEFYT RIPHDFGLRT PPLIRTQKEL SEKIQLLEAL GDIEIAIKLV KTELQSPEHP LDQHYRNLHC ALRPLDHESY EFKVISQYLQ STHAPTHSDY TMTLLDLFEV EKDGEKEAFR EDLHNRMLLW HGSRMSNWVG ILSHGLRIAP PEAPITGYMF GKGIYFADMS SKSANYCFAS RLKNTGLLLL SEVALGQCNE LLEANPKAEG LLQGKHSTKG LGKMAPSSAH FVTLNGSTVP LGPASDTGIL NPDGYTLNYN EYIVYNPNQV RMRYLLKVQF NFLQLW
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.