Learn More
Novus Biologicals™ Recombinant Human Park7/DJ-1 His Protein
Product Code. 18745353 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Product Code. 18745353
Brand: Novus Biologicals™ NBC118334
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-189 of Human Park7/DJ-1 The Recombinant Human Park7/DJ-1 Protein is derived from E. coli. The Recombinant Human Park7/DJ-1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.1 mg