Learn More
Novus Biologicals™ Recombinant Human NKG2E/KLRC3 His Protein
Product Code. 18711754 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Product Code. Quantity unitSize
18711754 0.1 mg 0.1mg
1 options
Product Code. 18711754 Supplier Novus Biologicals™ Supplier No. NBP223171
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 94-240 of Human NKG2E/KLRC3 The Recombinant Human NKG2E/KLRC3 Protein is derived from E. coli. The Recombinant Human NKG2E/KLRC3 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.1 mg