missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human MRPL28 His Protein

Product Code. 18140592 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
This item is not returnable. View return policy

Product Code. 18140592

Brand: Novus Biologicals™ NBP223241

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 56-256 of Human MRPL28 The Recombinant Human MRPL28 Protein is derived from E. coli. The Recombinant Human MRPL28 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_006419
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.5), 0.1 M NaCl, 10% glycerol, 1mM DTT
Gene ID (Entrez) 10573
Molecular Weight (g/mol) 25.8kDa
Name MRPL28 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen MRPL28, 56-256aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MNGQRERVED VPIPIYFPPE SQRGLWGGEG WILGQIYANN DKLSKRLKKV WKPQLFEREF YSEILDKKFT VTVTMRTLDL IDEAYGLDFY ILKTPKEDLC SKFGMDLKRG MLLRLARQDP QLHPEDPERR AAIYDKYKEF AIPEEEAEWV GLTLEEAIEK QRLLEEKDPV PLFKIYVAEL IQQLQQQALS EPAVVQKRAS GQ
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.