missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human MAD3 His Protein

Product Code. 18147851 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18147851

Brand: Novus Biologicals™ NBP223219

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-206 of Human MAD3 The Recombinant Human MAD3 Protein is derived from E. coli. The Recombinant Human MAD3 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_112590
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 83463
Molecular Weight (g/mol) 25.9kDa
Name MAD3 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen MXD3, 1-206aa Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMEPLASN IQVLLQAAEF LERREREAEH GYASLCPHRS PGPIHRRKKR PPQAPGAQDS GRSVHNELEK RRRAQLKRCL ERLKQQMPLG ADCARYTTLS LLRRARMHIQ KLEDQEQRAR QLKERLRSKQ QSLQRQLEQL RGLAGAAERE RLRADSLDSS GLSSERSDSD QEELEVDVES LVFGGEAELL RGFVAGQEHS YSHGGGAWL
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.