missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human SF20/MYDGF His Protein

Product Code. 18260243 Shop All Bio Techne Products
Click to view available options
:
0.1mg; Unlabeled
This item is not returnable. View return policy

Product Code. 18260243

Brand: Novus Biologicals™ NBC122603

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 33-173 of Human IL25 The Recombinant Human IL25 Protein is derived from E. coli. The Recombinant Human IL25 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_061980
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 20% glycerol
Gene ID (Entrez) 56005
Molecular Weight (g/mol) 18kDa
Name IL25 Protein
Purification Method Protein
Immunogen MGSSHHHHHH SSGLVPRGSH MSEPTTVAFD VRPGGVVHSF SHNVGPGDKY TCMFTYASQG GTNEQWQMSL GTSEDHQHFT CTIWRPQGKS YLYFTQFKAE VRGAEIEYAM AYSKAAFERE SDVPLKTEEF EVTKTAVAHR PGAFKAELSK LVIVAKASRT EL
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Gene Alias C19orf10, chromosome 19 open reading frame 10, EUROIMAGE1875335, IL25, IL27, IL27w, interleukin 27 working designation, interleukin-25, Ly6elg, MYDGF, myeloid-derived growth factor, R33729_1, SF20, Stromal cell-derived growth factor SF20, UPF0556 protein C19orf10
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Protein SF20/MYDGF
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.