missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human HMGB1/HMG-1 His Protein

Product Code. 18251024 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18251024 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18251024 Supplier Novus Biologicals™ Supplier No. NBC118488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-215 of Human HMGB1/HMG-1 The Recombinant Human HMGB1/HMG-1 Protein is derived from E. coli. The Recombinant Human HMGB1/HMG-1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09429
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 1 mM EDTA, 0.5 MM DTT, 10% glycerol
Gene ID (Entrez) 3146
Molecular Weight (g/mol) M.W. (theoretical): 25 kDa
Name HMGB1/HMG-1 Protein
Purification Method Protein
Quantity 0.1 mg
Source Baculovirus (Sf9 insect cells)
Immunogen HMGB1, 1-215 aa, Human, His tagged, Recombinant, Baculovirus (NP_002119).
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Regulatory Status RUO
Endotoxin Concentration <1.0 EU / 1 μg of protein (determined by LAL method)
Gene Symbol HMGB1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Chromatin Research, Neuroscience
Purity or Quality Grade >90%, by SDS-PAGE
Protein HMGB1/HMG-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.