missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Histone H2AX His Protein
Shop All Bio Techne Products
Click to view available options
:
0.01mg; Unlabeled
Description
Recombinant H2AX protein corresponding to amino acids 2 - 143. Source: E. coli (Accession #: P16104) The Recombinant Human Histone H2AX His Protein is derived from E. coli. The Recombinant Human Histone H2AX His Protein has been validated for the following applications: Western Blot, SDS-Page.
Specifications
Specifications
| For Use With (Application) | SDS-PAGE, Western Blot |
| Formulation | 50mM Tris pH 8.0, 200mM NaCl, 8 M Urea, 10mM Imidazole |
| Gene ID (Entrez) | 3014 |
| Name | Histone H2AX |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Gene Alias | H2A.X, H2A/X, H2AFX |
| Cross Reactivity | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction