missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human GPT2 His Protein

Product Code. 18167581 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
This item is not returnable. View return policy

Product Code. 18167581

Brand: Novus Biologicals™ NBP223077

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-523 of Human GPT2 The Recombinant Human GPT2 Protein is derived from E. coli. The Recombinant Human GPT2 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_597700
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 7.5), 30% glycerol, 0.2 M NaCl, 2mM DTT
Gene ID (Entrez) 84706
Molecular Weight (g/mol) 60.3kDa
Name GPT2 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen GPT2, 1-523aa Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMQRAAAL VRRGCGPRTP SSWGRSQSSA AAEASAVLKV RPERSRRERI LTLESMNPQV KAVEYAVRGP IVLKAGEIEL ELQRGIKKPF TEVIRANIGD AQAMGQQPIT FLRQVMALCT YPNLLDSPSF PEDAKKRARR ILQACGGNSL GSYSASQGVN CIREDVAAYI TRRDGGVPAD PDNIYLTTGA SDGISTILKI LVSGGGKSRT GVMIPIPQYP LYSAVISELD AIQVNYYLDE ENCWALNVNE LRRAVQEAKD HCDPKVLCII NPGNPTGQVQ SRKCIEDVIH FAWEEKLFLL ADEVYQDNVY SPDCRFHSFK KVLYEMGPEY SSNVELASFH STSKGYMGEC GYRGGYMEVI NLHPEIKGQL VKLLSVRLCP PVSGQAAMDI VVNPPVAGEE SFEQFSREKE SVLGNLAKKA KLTEDLFNQV PGIHCNPLQG AMYAFPRIFI PAKAVEAAQA HQMAPDMFYC MKLLEETGIC VVPGSGFGQR EGTYHFRMTI LPPVEKLKTV LQKVKDFHIN FLEKYA
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category metabolism
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.