missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Glutathione S Transferase kappa 1 Protein

Product Code. 18255814 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
This item is not returnable. View return policy

Product Code. 18255814

Brand: Novus Biologicals™ NBP137083

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-226 of Human Glutathione S Transferase kappa 1 The Recombinant Human Glutathione S Transferase kappa 1 Protein is derived from E. coli. The Recombinant Human Glutathione S Transferase kappa 1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_057001
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1 mM DTT
Gene ID (Entrez) 373156
Molecular Weight (g/mol) M.W. (theoretical): 25.5 kDa
Name Glutathione S Transferase kappa 1 Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MGPLPRTVEL FYDVLSPYSW LGFEILCRYQ NIWNINLQLR PSLITGIMKD SGNKPPGLLP RKGLYMANDL KLLRHHLQIP IHFPKDFLSV MLEKGSLSAM RFLTAVNLEH PEMLEKASRE LWMRVWSRNE DITEPQSILA AAEKAGMSAE QAQGLLEKIA TPKVKNQLKE TTEAACRYGA FGLPITVAHV DGQTHMLFGS DRMELLAHLL GEKWMGPIPP AVNARL
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Gene Symbol GSTK1
Cross Reactivity Human
Purity or Quality Grade >90%, by SDS-PAGE
Protein Glutathione S Transferase kappa 1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.