missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Cyclin I His Protein

Product Code. 18188741 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18188741

Brand: Novus Biologicals™ NBP222970

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-377 of Human Cyclin I The Recombinant Human Cyclin I Protein is derived from E. coli. The Recombinant Human Cyclin I Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_006826
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 10983
Molecular Weight (g/mol) 44.9kDa
Name Cyclin Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen CCN1, 1-377aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMKFPGPL ENQRLSFLLE KAITREAQMW KVNVRKMPSN QNVSPSQRDE VIQWLAKLKY QFNLYPETFA LASSLLDRFL ATVKAHPKYL SCIAISCFFL AAKTVEEDER IPVLKVLARD SFCGCSSSEI LRMERIILDK LNWDLHTATP LDFLHIFHAI AVSTRPQLLF SLPKLSPSQH LAVLTKQLLH CMACNQLLQF RGSMLALAMV SLEMEKLIPD WLSLTIELLQ KAQMDSSQLI HCRELVAHHL STLQSSLPLN SVYVYRPLKH TLVTCDKGVF RLHPSSVPGP DFSKDNSKPE VPVRGTAAFY HHLPAASGCK QTSTKRKVEE MEVDDFYDGI KRLYNEDNVS ENVGSVCGTD LSRQEGHASP CPPLQPVSVM
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.