missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Apolipoprotein L4 His Protein

Product Code. 18169041 Shop All Bio Techne Products
Click to view available options
Quantity:
0.02 mg
Unit Size:
0.02mg
This item is not returnable. View return policy

Product Code. 18169041

Brand: Novus Biologicals™ NBP222692

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-348 of Human Apolipoprotein L4 The Recombinant Human Apolipoprotein L4 Protein is derived from E. coli. The Recombinant Human Apolipoprotein L4 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_085146
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 80832
Molecular Weight (g/mol) 41.1kDa
Name Apolipoprotein L4 Protein
Purification Method SDS-PAGE
Quantity 0.02 mg
Source E. Coli
Immunogen APOL4, 1-348aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMGSWVQL ITSVGVQQNH PGWTVAGQFQ EKKRFTEEVI EYFQKKVSPV HLKILLTSDE AWKRFVRVAE LPREEADALY EALKNLTPYV AIEDKDMQQK EQQFREWFLK EFPQIRWKIQ ESIERLRVIA NEIEKVHRGC VIANVVSGST GILSVIGVML APFTAGLSLS ITAAGVGLGI ASATAGIASS IVENTYTRSA ELTASRLTAT STDQLEALRD ILRDITPNVL SFALDFDEAT KMIANDVHTL RRSKATVGRP LIAWRYVPIN VVETLRTRGA PTRIVRKVAR NLGKATSGVL VVLDVVNLVQ DSLDLHKGAK SESAESLRQW AQELEENLNE LTHIHQSLKA G
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category metabolism
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.