missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human AMSH/STAMBP His Protein

Product Code. 18196391 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18196391

Brand: Novus Biologicals™ NBP222825

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-424 of Human AMSH/STAMBP The Recombinant Human AMSH/STAMBP Protein is derived from E. coli. The Recombinant Human AMSH/STAMBP Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_998787
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 10% glycerol, 1mM DTT
Gene ID (Entrez) 10617
Molecular Weight (g/mol) theoretical_molecular_weight: L606
Name AMSH/STAMBP Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen STAMBP, 1-424aa Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMSDHGDV SLPPEDRVRA LSQLGSAVEV NEDIPPRRYF RSGVEIIRMA SIYSEEGNIE HAFILYNKYI TLFIEKLPKH RDYKSAVIPE KKDTVKKLKE IAFPKAEELK AELLKRYTKE YTEYNEEKKK EAEELARNMA IQQELEKEKQ RVAQQKQQQL EQEQFHAFEE MIRNQELEKE RLKIVQEFGK VDPGLGGPLV PDLEKPSLDV FPTLTVSSIQ PSDCHTTVRP AKPPVVDRSL KPGALSNSES IPTIDGLRHV VVPGRLCPQF LQLASANTAR GVETCGILCG KLMRNEFTIT HVLIPKQSAG SDYCNTENEE ELFLIQDQQG LITLGWIHTH PTQTAFLSSV DLHTHCSYQM MLPESVAIVC SPKFQETGFF KLTDHGLEEI SSCRQKGFHP HSKDPPLFCS CSHVTVVDRA VTITDLR
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.