missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Histamine H1R Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Description
A blocking peptide from human HRH1. Source: Synthetic Amino Acid Sequence: C-NENFKKTFKRILH The Histamine H1R Blocking Peptide is derived from Synthetic. The Histamine H1R Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Gene ID (Entrez) | 3269 |
| Species | Human |
| Purity | >50%, by HPLC |
| Accession Number | NP_000852.1 |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | Lyophilized from a 5 pmol/ul solution in 5% (v/v) acetonitrile/H2O and TFA salt. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB600-795. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | H1R, H1-R, hisH1, histamine receptor H1, subclass H1 |
| Gene Symbol | HRH1 |
| Product Type | Histamine H1 R |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction