missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ GMDS Protein

Product Code. 18215923 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
100 μg
250 μg
Formulation:
20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 2mM DTT
20mM Tris-HCl buffer (pH 8.0) containing 30% glycerol, 0.1M NaCl, 1mM DTT, 0.1mM PMSF
Unit Size:
0.5mg
100µg
250µg
This item is not returnable. View return policy

Product Code. 18215923

Brand: Novus Biologicals™ NBP172521100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-372 of Human GMDS The Recombinant Human GMDS Protein is derived from E. coli. The Recombinant Human GMDS Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 30% glycerol, 0.1M NaCl, 1mM DTT, 0.1mM PMSF
Molecular Weight (g/mol) 44.1kDa
Purification Method SDS-PAGE
Quantity 100 μg
Source Human
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name GMDS
Conjugate Unconjugated
Form Liquid
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.