missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Glycerol Kinase Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
A blocking peptide from human Glycerol Kinase. Source: Synthetic Amino Acid Sequence: (Accession #: NP_976325)MAAGAAEGVGVWSLEPEDLSAVTMERFEPQINAEESEIRYSTWKKAVMKS The Glycerol Kinase Blocking Peptide is derived from Synthetic. The Glycerol Kinase Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Gene ID (Entrez) | 2710 |
| Species | Human |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | Lyophilized with sterile distilled water. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-57033. This Synthetic Peptide is Designed for Use in an Antibody Competition Assay with Its Corresponding Antibody. Use of This Product in Any Other Assay Has Not Yet Been Tested. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | ATP:glycerol 3-phosphotransferase, EC 2.7.1.30, GK1, GKD, Glycerokinase, glycerol kinase |
| Gene Symbol | GK |
| Molecular Weight (g/mol) | 58kDa |
| Product Type | Glycerol Kinase |
| Quantity | 100 μg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction