missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Gene ID (Entrez) | 2740 |
| Species | Human |
| Accession Number | P43220 |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | PBS, pH 7.4 |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NLS1205. To Block The Specific Antibody Activity in Immunohistochemistry Analysis, Pre-Incubate The Antibody with a 10-100 x Molar Excess of Peptide for 60 Minutes (at Room Temperature. For Further Blocking Peptide Related Protocol, Click Here .) |
| Gene Alias | GLP-1 receptor, GLP-1 R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331 |
| Gene Symbol | GLP1R |
| Product Type | GLP-1R |
| Quantity | 0.05 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?