missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ gamma Tubulin Protein

Product Code. 18232812 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18232812

Brand: Novus Biologicals™ NBP172534100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-451 of Human gamma Tubulin The Recombinant Human gamma Tubulin Protein is derived from E. coli. The Recombinant Human gamma Tubulin Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.0mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) 1M Urea, 5% Glycerol.
Molecular Weight (g/mol) 53.3kDa
Purification Method SDS-PAGE
Quantity 100 μg
Source Recombinant human TUBG1 protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen TUBG1, 1-451 aa, Human, His tag N terminus E. coli
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name gamma Tubulin
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.