missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ eIF4E Protein

Product Code. 18273232 Shop All Bio Techne Products
Click to view available options
Quantity:
0.01 mg
0.05 mg
0.25 mg
Unit Size:
0.05mg
0.25mg
10µg
This item is not returnable. View return policy

Product Code. 18273232

Brand: Novus Biologicals™ NBP1453140.05MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-217 of Human eIF4E The Recombinant Human eIF4E Protein is derived from E. coli. The Recombinant Human eIF4E Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001959
Concentration 0.5mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation In 20mM Tris-HCl Buffer (pH 8.0) containing 10% Glycerol
Gene ID (Entrez) 1977
Molecular Weight (g/mol) 27.2kDa
Purification Method Protein
Quantity 0.05 mg
Source Recombinant human EIF4E protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen EIF4E, 1-217aa, Human, His tag, E. coli (NP_001959).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name eIF4E
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.