missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CTRP6 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Packungsgröße:
0.05mg
Beschreibung
A blocking peptide containing 15 amino acids near the center of human CTRP6. Source: Synthetic (Accession #: AAQ88740) The CTRP6 Blocking Peptide has been validated for the following applications: Antibody Competition.
Spezifikation
Spezifikation
| Gene ID (Entrez) | 114904 |
| Species | Human |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-76633. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | C1q and tumor necrosis factor related protein 6, complement C1q tumor necrosis factor-related protein 6, complement-c1q tumor necrosis factor-related protein 6, CTRP6ZACRP6 |
| Gene Symbol | C1QTNF6 |
| Product Type | CTRP6 |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur