missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CCL2/MCP1 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
Description
A peptide to CCL2.
Specifications
Specifications
| Gene ID (Entrez) | 6347 |
| Species | Human |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-07034. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | C-C motif chemokine 2, chemokine (C-C motif) ligand 2, GDCF-2, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, MCP1 Monocyte chemotactic protein 1, MCP-1 Small-inducible cytokine A2, MGC9434, Monocyte chemoattractant protein 1, monocyte chemoattractant protein-1, SCYA2HSMCR30, small inducible cytokine A2 (monocyte chemotactic protein 1, homologous tomouse Sig-je), small inducible cytokine subfamily A (Cys-Cys), member 2, SMC-CF |
| Gene Symbol | CCL2 |
| Product Type | CCL2/MCP1 |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction