missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ BID Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
:
0.1 mg
Description
The BID Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC27261 and based on accession number NM_1196. The protein contains a C-terminal DDK tag. |
| For Use With (Application) | Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens BH3 interacting domain death agonist (BID), transcript variant 2, mRNA. |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Research Category | Apoptosis, Cancer, Tumor Suppressors |
| Target | BID |
| Synonym | apoptic death agonist, BH3 interacting domain death agonist, BH3-interacting domain death agonist, BID isoform ES(1 b), BID isoform L(2), BID isoform Si6, desmocollin type 4, FP497, Human BID coding sequence, MGC15319, MGC42355, p22 BID, tbid |
| Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction