missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ BACE-1 Antibody Blocking Peptide

Product Code. 18277455 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
This item is not returnable. View return policy

Product Code. 18277455

Brand: Novus Biologicals™ NBP177307PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide containing 17 amino acids near the carboxy terminus of human BACE. Source: Synthetic (Accession #: AF190725) The BACE-1 Blocking Peptide has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 23621
Species Human
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-77307. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias APP beta-secretase, ASP2, Aspartyl protease 2, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2
Gene Symbol BACE1
Product Type BACE-1
Quantity 0.05 mg
Regulatory Status RUO - research use only
Source Synthetic
Termination 17 amino acids near the carboxy terminus of human BACE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.