missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ APO-BRDU (TUNEL) Apoptosis Kit

Product Code. 18735533 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18735533

Brand: Novus Biologicals™ NBP231161

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Apoptosis Kit

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 1 Kit
Kit Contents Negative Control Cells, Wash Buffer, Reaction Buffer, TdT Enzyme, Br-dUTP, Rinsing Buffer, Propidium Iodide/RNase DNA staining solution, Fluorescein-anti-BrdU antibody
For Use With (Application) Flow Cytometry, Immunocytochemistry/Immunofluorescence
Product Type APO-BRDU (TUNEL) Apoptosis Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.