missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Apc10 Antibody Blocking Peptide

Product Code. 18298895 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18298895 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18298895 Supplier Novus Biologicals™ Supplier No. NBP177157PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide containing 16 amino acids near the center of human APC10. Source: Synthetic (Accession #: EAX05052) The Apc10 Blocking Peptide has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 10393
Species Human
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-77157. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias anaphase promoting complex subunit 10, anaphase-promoting complex subunit 10, APC10DOC1, Cyclosome subunit 10, DKFZP564L0562
Gene Symbol ANAPC10
Product Type Apc10
Quantity 0.05 mg
Regulatory Status RUO - research use only
Source Synthetic
Termination 16 amino acids near the center of human APC10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.