missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ AP2 alpha Antibody Blocking Peptide

Product Code. 18016476 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
This item is not returnable. View return policy

Product Code. 18016476

Brand: Novus Biologicals™ NBP196941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide from AP2. Source: Synthetic The AP2 alpha Blocking Peptide has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 7020
Species Human
Purity >95%
Accession Number O95782
Content And Storage Store at -20 C short term. Aliquot and store (at -80 C long term. Avoid freeze-thaw cycles.)
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
For Use With (Application) This product should be used at 1.0μg per 1.0μL of antiserum in per assay.
Gene Alias Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2 alpha, AP-2 transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha
Gene Symbol TFAP2A
Product Type AP2 alpha Peptide
Quantity 0.05 mg
Regulatory Status RUO - research use only
Source Synthetic
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.