missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ABCG1 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
Description
A peptide to ABCG1.
Specifications
Specifications
| Gene ID (Entrez) | 9619 |
| Species | Human |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB 400-132. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | ABC transporter 8, ABC8ATP-binding cassette sub-family G member 1, ATP-binding cassette transporter 8, ATP-binding cassette, sub-family G (WHITE), member 1, homolog of Drosophila white, MGC34313, White protein homolog, white protein homolog (ATP-binding cassette transporter 8), 10 ATP-binding cassette transporter member 1 of subfamily G, WHITE1, WHT1 |
| Gene Symbol | ABCG1 |
| Product Type | ABCG1 |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction