missing translation for 'onlineSavingsMsg'
Learn More

Novobiocin sodium salt, Thermo Scientific Chemicals

Catalog Number p-7058194
Click to view available options
Quantity:
5 g
1476-53-5
C31H35N2NaO11
634.61
MFCD00066541,MFCD00066541
AXOUUAINTJNFRS-UHFFFAOYNA-N
131673945
sodium 7-{[4-(carbamoyloxy)-3-hydroxy-5-methoxy-6,6-dimethyloxan-2-yl]oxy}-3-[4-hydroxy-3-(3-methylbut-2-en-1-yl)benzamido]-8-methyl-2,4-dioxo-3,4-dihydro-2H-1-benzopyran-3-ide
[Na+].COC1C(OC(N)=O)C(O)C(OC2=CC=C3C(=O)[C-](NC(=O)C4=CC=C(O)C(CC=C(C)C)=C4)C(=O)OC3=C2C)OC1(C)C
1476-53-5
C31H35N2NaO11
634.61
MFCD00066541,MFCD00066541
AXOUUAINTJNFRS-UHFFFAOYNA-N
131673945
sodium 7-{[4-(carbamoyloxy)-3-hydroxy-5-methoxy-6,6-dimethyloxan-2-yl]oxy}-3-[4-hydroxy-3-(3-methylbut-2-en-1-yl)benzamido]-8-methyl-2,4-dioxo-3,4-dihydro-2H-1-benzopyran-3-ide
[Na+].COC1C(OC(N)=O)C(O)C(OC2=CC=C3C(=O)[C-](NC(=O)C4=CC=C(O)C(CC=C(C)C)=C4)C(=O)OC3=C2C)OC1(C)C

Novobiocin sodium salt, CAS # 1476-53-5, is a topoisomerase II inhibitor capable of inhibiting DNA synthesis. This compound is produced by Streptomyces nieveus and shows antibiotic activity.

Topoisomerase II inhibitorNovobiocin sodium salt is effective an antistaphylococcal agent used in the treatment of Methicillin-resistant Staphylococcus aureus (MRSA). It is also used to inhibit the activity of retroviral RNA-dependent DNA polymerase as well as inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Novobiocin sodium salt, first isolated from Streptomyces nieveus, is a topoisomerase II inhibitor. It is a coumarin compound showing antibiotic activity
• Novobiocin sodium salt can inhibit DNA synthesis by inhibiting the topoisomerase II enzyme. Its antibacterial activity arises from competitive inhibition of the ATPase reaction catalyzed by the GyrB subunit

Application

• Novobiocin sodium salt is used to inhibit the activity of retroviral RNA-dependent DNA polymerase in laboratory experiments
• It is also used as an inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase.
• Novobiocin sodium salt shows anti-staphylococcal activity against methicillin-resistant Staphylococcus aureus, a gram-positive bacterium
• It permits the relaxation of negative DNA supercoils and has been used to generate positively supercoiled DNA
• This compound inhibits the chaperone activity of Hsp90

TRUSTED_SUSTAINABILITY
Type Novobiocin Sodium Salt
Physical Form Powder
Melting Point 215°C
Beilstein 3892910
Merck Index 14,6722
Solubility Information Soluble in water at 100mg/ml; Also soluble in ethanol or DMSOSoluble in water,ethanol,dimethyldulfoxide and methanol.
Formula Weight 634.61
Color Brown to White
Quantity 5 g
Chemical Name or Material Novobiocin Sodium Salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'