missing translation for 'onlineSavingsMsg'
Learn More

Novobiocin sodium, MedChemExpress

Product Code. 16412136
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16412136

Brand: Medchem Express HYB0425A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Novobiocin (Albamycin) sodium is a potent and orally active antibiotic. Novobiocin sodium also is a DNA gyrase inhibitor and a heat shock protein 90 (Hsp90) antagonist. Novobiocin sodium has the potential for the research of highly beta-lactam-resistant pneumococcal infections. Novobiocin sodium shows anti-orthopoxvirus activity.

Novobiocin (Albamycin) sodium is a potent and orally active antibiotic. Novobiocin sodium also is a DNA gyrase inhibitor and a heat shock protein 90 (Hsp90) antagonist. Novobiocin sodium has the potential for the research of highly beta-lactam-resistant pneumococcal infections. Novobiocin sodium shows anti-orthopoxvirus activity.

Specifications

Chemical Name or Material Novobiocin sodium
Physical Form Powder
CAS 1476-53-5
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C31H35N2NaO11
For Use With (Application) COVID-19-immunoregulation
Synonym Albamycin sodiumCathomycin sodium
Solubility Information H2O : 50 mg/mL (78.79 mM; Need ultrasonic) ∣DMSO : ≥ 30 mg/mL (47.27 mM)
SMILES CC1=C(OC2=O)C(C(O)=C2NC(C3=CC(C/C=C(C)/C)=C(O[Na])C=C3)=O)=CC=C1O[C@H](OC(C)(C)[C@H](OC)[C@H]4OC(N)=O)[C@@H]4O
Molecular Weight (g/mol) 634.61
Formula Weight 634.61
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.