missing translation for 'onlineSavingsMsg'
Learn More

Melford Novobiocin

Product Code. 30018580
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
Product Code. Quantity unitSize
30018580 25 g 25g
30018579 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 30018580

Brand: Melford N3020025.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Novobiocin Sodium Salt is an antibiotic that is a member of the coumarin class of antibiotics and is mainly effective against Gram-positive bacteria.

Novobiocin Sodium Salt is an antibiotic that is a member of the coumarin class of antibiotics and is mainly effective against Gram-positive bacteria. In microbiology and molecular biology research, Novobiocin Sodium Salt is used as a selective agent. It inhibits the activity of DNA gyrase, an enzyme involved in DNA replication and repair in bacteria.

Researchers can incorporate Novobiocin Sodium Salt into growth media to selectively allow the growth of cells or organisms that are resistant to Novobiocin, typically due to the presence of specific resistance genes. This chemical has also found applications in studying DNA topology, replication, and transcription in bacteria, making it valuable in molecular biology and genetic research.

Specifications

Chemical Name or Material Novobiocin
CAS 1476-53-5
Quantity 25 g
Molecular Formula C31H35N2NaO11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.