Learn More
Melford Novobiocin
Description
Novobiocin Sodium Salt is an antibiotic that is a member of the coumarin class of antibiotics and is mainly effective against Gram-positive bacteria.
Researchers can incorporate Novobiocin Sodium Salt into growth media to selectively allow the growth of cells or organisms that are resistant to Novobiocin, typically due to the presence of specific resistance genes. This chemical has also found applications in studying DNA topology, replication, and transcription in bacteria, making it valuable in molecular biology and genetic research.
Specifications
Specifications
| Chemical Name or Material | Novobiocin |
| CAS | 1476-53-5 |
| Quantity | 1 g |
| Molecular Formula | C31H35N2NaO11 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.