missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZOOM™ IPG Strips, pH 3-10L

Product Code. 12070216
Change view
Click to view available options
Quantity:
12 strips
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12070216 12 strips Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 12070216 Supplier Invitrogen™ Supplier No. ZM0018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
For Research Use Only. All usage must comply with product instructions.
 

Requires

ZOOM IPGRunner System

Oil-free, trouble-free 2D electrophoresis in a mini-gel format

  • First dimension separation, isoelectric focusing (IEF), is complete in less than three hours
  • The mini-gel design of the system is easy to handle, eliminates mineral oil overlays, and allows processing of up to 12 samples at once for high-throughput usage
  • Strips are 7cm long and contain a thin layer of polyacrylamide gel that includes a fixed pH gradient
  • Each strip is clearly labeled with a unique identifying number, pH range, and orientation marks
  • Strips are supplied attached to a tri-fold card for easy access and removal

2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The ZOOM™ IPGRunner™ Mini-Cell includes the ZOOM™ IPGRunner™ Buffer Core and Lid, Gel Tension Wedge, Buffer Dam, and the Mini- Cell chamber. The ZOOM™ IPGRunner™ Combo Kit includes the ZOOM™ IPGRunner™ Mini-Cell, electrode wicks, sealing tape, 10 ZOOM™ IPGRunner™ Cassettes, and 12 ZOOM™ Strips, pH 3-10 NL. The ZOOM™ IPGRunner™ Retrofit Kit for XCell SureLock™ Mini-Cell includes the ZOOM™ IPGRunner™ Buffer Core and Lid. The ZOOM™ IPGRunner™ Cassettes include electrode wicks and sealing tape.
Gel Size Mini
pH Range 3.0 - 10.0 (linear)
Quantity 12 strips
Shipping Condition Dry Ice
Product Line ZOOM
Product Type IPG Strip
For Use With (Equipment) XCell SureLock Mini-Cell

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.