missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Tris-Glycine ZOOM™ Mini Protein Gel, 4 to 20%, 1.0 mm, IPG-well

Product Code. 12035685
Change view
Click to view available options
Quantity:
10 gels (1 box)
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12035685 10 gels (1 box) Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 12035685 Supplier Invitrogen™ Supplier No. EC60261BOX

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

One box of 10 gels

Separation Range: 20 to 200 kd; Separation Type: Denaturing; For Use With: ZOOM IPGRunner Mini

  • Used to separate proteins based upon their molecular weight after isoelectric focusing (IEF)
  • 2D gels are generally designed to accept IEF tube gels, sample lanes sliced from vertical or horizontal IEF slab gels, or immobilized pH gradient (IPG) strips
  • Gels are usually run under denaturing conditions using SDS in the gel running buffers
  • The IPG well accommodates the full length of a 7cm IPG strip
  • In addition to the IPG well, ZOOM Gels have a separate well designed for use with a molecular weight standard
  • Standard 10cm x 10cm cassette format

Sample Loading Volume: 7cm strips; IPG Well

2D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage One box contains 10 gels. Store in refrigerator (2–8°C). Do not freeze. Shelf life is 8 weeks.
Gel Percentage 4 to 20%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Tris-Glycine
Sample Loading Volume 7 cm strips
Separation of Protein
Separation Range 20 to 200 kDa
Separation Type Denaturing
Wells 1-well
Mode of Separation Molecular Weight
Quantity 10 gels (1 box)
Shipping Condition Wet Ice
Thickness 1.0 mm
Well Design IPG Well
Length (Metric) 8 cm
Length (Metric) Gel 8 cm
Width (Metric) 8 cm
Width (Metric) Gel 8 cm
Product Line Novex, ZOOM
Product Type Gradient Page
For Use With (Equipment) ZOOM IPGRunner Mini
System Type ZOOM
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.