missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WesternBreeze™ Chemiluminescent Kit, anti-goat

Product Code. 10164612
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10164612 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10164612 Supplier Invitrogen™ Supplier No. WB7108

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Detect proteins that have been immobilized on membranes (nitrocellulose or PVDF) following western transfer or bound directly from solution (dot blots)

Detection is accomplished with a ready-to-use CDP-Star™ chemiluminescent substrate for alkaline phosphatase. Protein bands can be captured either by X-ray film or CDP-Star™ compatible imaging system.

WesternBreeze™ Chemiluminescent Kit offers:

  • High specificity, clean background
  • Ultra-sensitivity-femtogram levels detectable
  • Long-lasting signals-up to 5 days
  • Results in less than 3 hours
  • Permanent photographic image

Proteins, Expression, Isolation and Analysis, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Product Type Western Blot Kit
Content And Storage The WesternBreeze™ Chemiluminescent Kits include blocking solutions, primary antibody diluent, ready-to-use secondary antibody solution (anti-mouse, anti-rabbit or anti-goat), ready-to-use chemiluminescent substrate, wash solutions, incubation trays, pre-cut filter papers, polyester sheet for even substrate development on the membrane. Each kit contains complete reagents for 20 blots. Store the kits at +4°C. Guaranteed stable for 6 months when properly stored.
Reactivity Goat
Shipping Condition Wet Ice
Substrate Type AP (Alkaline Phosphatase) Substrate
Detection Method Chemiluminescence
Membrane Compatibility Nitrocellulose, PVDF
Product Line WesternBreeze
Cross Reactivity Goat
Quantity 1 kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.