missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex TNF- Mouse ELISA Kit

Product Code. 10314073
Change view
Click to view available options
Quantity:
192 Assays
96 Assays
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10314073 96 Assays Each
10272923 192 Assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10314073 Supplier Invitrogen™ Supplier No. KMC3011

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

Designed to detect and quantify the level of mouse TNF-α in serum, plasma, and supernatant using 96-well plates and a microplate reader

  • The total assay incubation time is only 2.5 hour
  • A monoclonal capture antibody specific for Mouse TNF-α has been coated onto the wells of the 96-well plate provided
  • Samples, including a standard of known Mouse TNF-α content, controls, and unknowns, are pipetted into these wells followed by an addition of a biotinylated polyclonal detector antibody
  • During the first incubation, Mouse TNF-α antigen binds to the immobilized capture antibody and to the solution phase biotinylated detector antibody on a second site
  • After washing, a streptavidin-horseradish peroxidase (HRP) is added
  • This binds to the biotinylated detection antibody to complete the four–member sandwich
  • After a second incubation and washing to remove all unbound enzyme, a substrate solution (TMB) is added, which is acted upon by the bound enzyme to produce color
  • The intensity of this colored product is directly proportional to the concentration of Mouse TNF-α present in the original specimen and the optical density can be read on a standard microplate reader

The kit comes with a capture antibody precoated to the plate, eliminating an overnight coating process. The plates are machine coated to provide low variability well-to-well. Furthermore, the plate comes as removable 8-well strips allowing you to run as few samples as you want. Each ELISA kit is validated for sensitivity, specificity, precision, and lot-to-lot consistency.

Apoptosis, Apoptosis Related Factors, Cell Analysis, Cell Signaling, Cell Viability, Proliferation and Function, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Sample Type Plasma, Serum, Supernatant
For Use With (Application) Colorimetric
Gene ID (Entrez) 21926
Kit Contents Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer
Precision <3pg/mL
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Target Tumor necrosis factor
Test Time 2.5 hr.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.