Learn More
Invitrogen™ Novex TNF- Mouse ELISA Kit
Description
Includes
Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
- The total assay incubation time is only 2.5 hour
- A monoclonal capture antibody specific for Mouse TNF-α has been coated onto the wells of the 96-well plate provided
- Samples, including a standard of known Mouse TNF-α content, controls, and unknowns, are pipetted into these wells followed by an addition of a biotinylated polyclonal detector antibody
- During the first incubation, Mouse TNF-α antigen binds to the immobilized capture antibody and to the solution phase biotinylated detector antibody on a second site
- After washing, a streptavidin-horseradish peroxidase (HRP) is added
- This binds to the biotinylated detection antibody to complete the four–member sandwich
- After a second incubation and washing to remove all unbound enzyme, a substrate solution (TMB) is added, which is acted upon by the bound enzyme to produce color
- The intensity of this colored product is directly proportional to the concentration of Mouse TNF-α present in the original specimen and the optical density can be read on a standard microplate reader
The kit comes with a capture antibody precoated to the plate, eliminating an overnight coating process. The plates are machine coated to provide low variability well-to-well. Furthermore, the plate comes as removable 8-well strips allowing you to run as few samples as you want. Each ELISA kit is validated for sensitivity, specificity, precision, and lot-to-lot consistency.
Apoptosis, Apoptosis Related Factors, Cell Analysis, Cell Signaling, Cell Viability, Proliferation and Function, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Wet ice
Specifications
Specifications
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Application) | Colorimetric |
| Gene ID (Entrez) | 21926 |
| Kit Contents | Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer |
| Precision | <3pg/mL |
| Quantity | 192 Assays |
| Regulatory Status | For research use only (RUO) |
| Target | Tumor necrosis factor |
| Test Time | 2.5 hr. |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.