Learn More
Invitrogen™ Novex TNF- Monkey ELISA Kit
Description
Includes
Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
TNF-α is produced by macrophages and has many immune effects.
Apoptosis, Apoptosis Related Factors, Cell Analysis, Cell Signaling, Cell Viability, Proliferation and Function, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Wet ice
Specifications
Specifications
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Application) | Colorimetric |
| Gene ID (Entrez) | 7124 |
| Kit Contents | Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer |
| Precision | <2pg/mL |
| Quantity | 96 Assays |
| Regulatory Status | For research use only (RUO) |
| Target | Tumor necrosis factor |
| Test Time | 4 hr. |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.