missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex TNF- Monkey ELISA Kit

Product Code. 10384633
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
192 Assays
96 Assays
missing translation for 'unitSize'
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10384633 192 Assays Each
10252583 96 Assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10384633 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' KPC3012

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

Quantifies natural and recombinant monkey TNF-α in serum, plasma, buffered solution, or cell culture medium

TNF-α is produced by macrophages and has many immune effects.

Apoptosis, Apoptosis Related Factors, Cell Analysis, Cell Signaling, Cell Viability, Proliferation and Function, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Sample Type Plasma, Serum, Supernatant
For Use With (Application) Colorimetric
Gene ID (Entrez) 7124
Kit Contents Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer
Precision <2pg/mL
Quantity 192 Assays
Regulatory Status For research use only (RUO)
Target Tumor necrosis factor
Test Time 4 hr.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.